4-Ethyl-5-methylthiophene-3-carbohydrazide
Supplier: Matrix Scientific
029837-500MG
101828-954EA
217.35
USD
101828-954
4-Ethyl-5-methylthiophene-3-carbohydrazide
4-Ethyl-5-methylthiophene-3-carbohydrazide
Hazard: Irritant
Formula:
C₈H₁₂N₂OS |
MDL Number:
MFCD03419844 CAS Number: 438225-41-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...