4,5-Dimethylthiophene-3-carbohydrazide

Supplier: Matrix Scientific

029836-500MG
101828-952EA 290.77 USD
101828-952
4,5-Dimethylthiophene-3-carbohydrazide
4,5-Dimethylthiophene-3-carbohydrazide

Hazard: Irritant

Formula: C₇H₁₀N₂OS
MDL Number: MFCD02245153
CAS Number: 524934-36-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR