5-Methylthiophene-3-carbohydrazide

Supplier: Matrix Scientific

029835-500MG
101828-950EA 213.85 USD
101828-950
5-Methylthiophene-3-carbohydrazide
5-Methylthiophene-3-carbohydrazide

Hazard: Irritant

Formula: C₆H₈N₂OS
MDL Number: MFCD01627991
CAS Number: 524731-02-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR