6-Bromo-2-(4-hydroxyphenyl)quinoline-4-carbohydrazide
Supplier: Matrix Scientific
029833-500MG
101828-946EA
290.77
USD
101828-946
6-Bromo-2-(4-hydroxyphenyl)quinoline-4-carbohydrazide
6-Bromo-2-(4-hydroxyphenyl)quinoline-4-carbohydrazide
Hazard: Irritant
Formula:
C₁₆H₁₂BrN₃O₂ |
MDL Number:
MFCD00814755 CAS Number: 351327-31-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...