4-(1H-Pyrrol-1-yl)benzohydrazide

Supplier: Matrix Scientific

029826-500MG
101828-934EA 144.9 USD
101828-934
4-(1H-Pyrrol-1-yl)benzohydrazide
4-(1H-Pyrrol-1-yl)benzohydrazide

Hazard: Irritant

Formula: C₁₁H₁₁N₃O
MDL Number: MFCD02194575
CAS Number: 112575-84-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR