4-(Cyclopentyloxy)benzohydrazide

Supplier: Matrix Scientific

029825-500MG
101828-932EA 336.92 USD
101828-932
4-(Cyclopentyloxy)benzohydrazide
4-(Cyclopentyloxy)benzohydrazide

Hazard: Irritant

Formula: C₁₂H₁₆N₂O₂
MDL Number: MFCD03423142
CAS Number: 438213-35-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR