2,3-Dichlorobenzhydrazide
Supplier: Matrix Scientific
Synonyms:
2,3-Dichlorobenzohydrazide
029824-500MG
101828-930EA
100
USD
101828-930
2,3-Dichlorobenzhydrazide
2,3-Dichlorobenzhydrazide
Hazard: Irritant
Formula:
C₇H₆Cl₂N₂O MW: 205.0400 g/mol |
MDL Number:
MFCD02612553 CAS Number: 438197-19-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...