2-(1,3-Benzodioxol-5-yl)quinoline-4-carbohydrazide

Supplier: Matrix Scientific

029819-500MG
101828-920EA 217.35 USD
101828-920
2-(1,3-Benzodioxol-5-yl)quinoline-4-carbohydrazide
2-(1,3-Benzodioxol-5-yl)quinoline-4-carbohydrazide

Hazard: Irritant

Formula: C₁₇H₁₃N₃O₃
MDL Number: MFCD02244464
CAS Number: 524932-89-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR