2-(5-Methylthien-2-yl)quinoline-4-carbohydrazide

Supplier: Matrix Scientific

029816-500MG
101828-914EA 290.77 USD
101828-914
2-(5-Methylthien-2-yl)quinoline-4-carbohydrazide
2-(5-Methylthien-2-yl)quinoline-4-carbohydrazide

Hazard: Irritant

Formula: C₁₅H₁₃N₃OS
MDL Number: MFCD00814786
CAS Number: 351327-53-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR