2-Pyridin-2-ylquinoline-4-carbohydrazide

Supplier: Matrix Scientific

029814-500MG
101828-910EA 290.77 USD
101828-910
2-Pyridin-2-ylquinoline-4-carbohydrazide
2-Pyridin-2-ylquinoline-4-carbohydrazide

Hazard: Irritant

Formula: C₁₅H₁₂N₄O
MDL Number: MFCD02090830
CAS Number: 5109-96-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR