2-(4-Bromophenyl)quinoline-4-carbohydrazide

Supplier: Matrix Scientific

029810-500MG
101828-902EA 290.77 USD
101828-902
2-(4-Bromophenyl)quinoline-4-carbohydrazide
2-(4-Bromophenyl)quinoline-4-carbohydrazide

Hazard: Irritant

Formula: C₁₆H₁₂BrN₃O
MDL Number: MFCD01421812
CAS Number: 351899-02-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR