2-(3-Chlorophenyl)quinoline-4-carbohydrazide

Supplier: Matrix Scientific

029806-500MG
101828-894EA 290.77 USD
101828-894
2-(3-Chlorophenyl)quinoline-4-carbohydrazide
2-(3-Chlorophenyl)quinoline-4-carbohydrazide

Hazard: Irritant

Formula: C₁₆H₁₂ClN₃O
MDL Number: MFCD03420728
CAS Number: 886628-78-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR