2-(4-Chlorophenyl)quinoline-4-carbohydrazide

Supplier: Matrix Scientific

029805-500MG
101828-892EA 217.35 USD
101828-892
2-(4-Chlorophenyl)quinoline-4-carbohydrazide
2-(4-Chlorophenyl)quinoline-4-carbohydrazide

Hazard: Irritant

Formula: C₁₆H₁₂ClN₃O
MDL Number: MFCD01471407
CAS Number: 884837-13-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR