2-(3-Propoxyphenyl)quinoline-4-carbohydrazide
Supplier: Matrix Scientific
029797-500MG
101828-876EA
290.77
USD
101828-876
2-(3-Propoxyphenyl)quinoline-4-carbohydrazide
2-(3-Propoxyphenyl)quinoline-4-carbohydrazide
Hazard: Irritant
Formula:
C₁₉H₁₉N₃O₂ |
MDL Number:
MFCD03420724 CAS Number: 438228-97-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...