2-(4-Propoxyphenyl)quinoline-4-carbohydrazide

Supplier: Matrix Scientific

029796-500MG
101828-874EA 290.77 USD
101828-874
2-(4-Propoxyphenyl)quinoline-4-carbohydrazide
2-(4-Propoxyphenyl)quinoline-4-carbohydrazide

Hazard: Irritant

Formula: C₁₉H₁₉N₃O₂
MDL Number: MFCD02375215
CAS Number: 51842-79-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR