2-(2,4-Diethoxyphenyl)quinoline-4-carbohydrazide
Supplier: Matrix Scientific
029794-500MG
101828-870EA
290.77
USD
101828-870
2-(2,4-Diethoxyphenyl)quinoline-4-carbohydrazide
2-(2,4-Diethoxyphenyl)quinoline-4-carbohydrazide
Hazard: Irritant
Formula:
C₂₀H₂₁N₃O₃ |
MDL Number:
MFCD03420723 CAS Number: 906789-68-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...