2-(4-tert-Butylphenyl)quinoline-4-carbohydrazide

Supplier: Matrix Scientific

029793-500MG
101828-868EA 217.35 USD
101828-868
2-(4-tert-Butylphenyl)quinoline-4-carbohydrazide
2-(4-tert-Butylphenyl)quinoline-4-carbohydrazide

Hazard: Irritant

Formula: C₂₀H₂₁N₃O
MDL Number: MFCD02244449
CAS Number: 524933-35-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR