2-(2-Ethoxyphenyl)quinoline-4-carbohydrazide

Supplier: Matrix Scientific

029792-500MG
101828-866EA 217.35 USD
101828-866
2-(2-Ethoxyphenyl)quinoline-4-carbohydrazide
2-(2-Ethoxyphenyl)quinoline-4-carbohydrazide

Hazard: Irritant

Formula: C₁₈H₁₇N₃O₂
MDL Number: MFCD02375051
CAS Number: 524932-37-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR