2-(3-Ethoxyphenyl)quinoline-4-carbohydrazide

Supplier: Matrix Scientific

029791-500MG
101828-864EA 290.77 USD
101828-864
2-(3-Ethoxyphenyl)quinoline-4-carbohydrazide
2-(3-Ethoxyphenyl)quinoline-4-carbohydrazide

Hazard: Irritant

Formula: C₁₈H₁₇N₃O₂
MDL Number: MFCD02244500
CAS Number: 524932-92-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR