2-(3-Methoxyphenyl)quinoline-4-carbohydrazide

Supplier: Matrix Scientific

029788-500MG
101828-858EA 290.77 USD
101828-858
2-(3-Methoxyphenyl)quinoline-4-carbohydrazide
2-(3-Methoxyphenyl)quinoline-4-carbohydrazide

Hazard: Irritant

Formula: C₁₇H₁₅N₃O₂
MDL Number: MFCD02373523
CAS Number: 524927-22-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR