2-(4-Isobutylphenyl)quinoline-4-carbohydrazide

Supplier: Matrix Scientific

029786-500MG
101828-854EA 290.77 USD
101828-854
2-(4-Isobutylphenyl)quinoline-4-carbohydrazide
2-(4-Isobutylphenyl)quinoline-4-carbohydrazide

Hazard: Irritant

Formula: C₂₀H₂₁N₃O
MDL Number: MFCD03420722
CAS Number: 588680-37-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR