2-(4-Isopropylphenyl)quinoline-4-carbohydrazide

Supplier: Matrix Scientific

029785-500MG
101828-852EA 217.35 USD
101828-852
2-(4-Isopropylphenyl)quinoline-4-carbohydrazide
2-(4-Isopropylphenyl)quinoline-4-carbohydrazide

Hazard: Irritant

Formula: C₁₉H₁₉N₃O
MDL Number: MFCD02176430
CAS Number: 402481-93-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR