2-(4-Ethylphenyl)quinoline-4-carbohydrazide

Supplier: Matrix Scientific

029784-500MG
101828-850EA 290.77 USD
101828-850
2-(4-Ethylphenyl)quinoline-4-carbohydrazide
2-(4-Ethylphenyl)quinoline-4-carbohydrazide

Hazard: Irritant

Formula: C₁₈H₁₇N₃O
MDL Number: MFCD02609415
CAS Number: 524733-42-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR