2-(2,5-Dimethylphenyl)quinoline-4-carbohydrazide

Supplier: Matrix Scientific

029783-500MG
101828-848EA 290.77 USD
101828-848
2-(2,5-Dimethylphenyl)quinoline-4-carbohydrazide
2-(2,5-Dimethylphenyl)quinoline-4-carbohydrazide

Hazard: Irritant

Formula: C₁₈H₁₇N₃O
MDL Number: MFCD01993856
CAS Number: 351000-24-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR