2-(2,4-Dimethylphenyl)quinoline-4-carbohydrazide

Supplier: Matrix Scientific

029781-500MG
101828-844EA 290.77 USD
101828-844
2-(2,4-Dimethylphenyl)quinoline-4-carbohydrazide
2-(2,4-Dimethylphenyl)quinoline-4-carbohydrazide

Hazard: Irritant

Formula: C₁₈H₁₇N₃O
MDL Number: MFCD02244921
CAS Number: 524934-05-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR