2-(2-Methylphenyl)quinoline-4-carbohydrazide

Supplier: Matrix Scientific

029780-500MG
101828-842EA 290.77 USD
101828-842
2-(2-Methylphenyl)quinoline-4-carbohydrazide
2-(2-Methylphenyl)quinoline-4-carbohydrazide

Hazard: Irritant

Formula: C₁₇H₁₅N₃O
MDL Number: MFCD02612351
CAS Number: 438197-10-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR