2-[4-(1-Methyl-1-phenylethyl)phenoxy]propanohydrazide
Supplier: Matrix Scientific
029771-500MG
101828-824EA
146.15
USD
101828-824
2-[4-(1-Methyl-1-phenylethyl)phenoxy]propanohydrazide
2-[4-(1-Methyl-1-phenylethyl)phenoxy]propanohydrazide
Hazard: Irritant
Formula:
C₁₈H₂₂N₂O₂ MW: 298.39 g/mol |
MDL Number:
MFCD03423136 CAS Number: 70757-69-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...