2-(2-Chloro-4-fluorophenoxy)propanohydrazide

Supplier: Matrix Scientific

029770-500MG
101828-822EA 109.25 USD
101828-822
2-(2-Chloro-4-fluorophenoxy)propanohydrazide
2-(2-Chloro-4-fluorophenoxy)propanohydrazide

Hazard: Irritant

Formula: C₉H₁₀ClFN₂O₂
MDL Number: MFCD03423135
CAS Number: 588677-35-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR