2-(4-Fluorophenoxy)propanohydrazide

Supplier: Matrix Scientific

029769-500MG
101828-820EA 109.25 USD
101828-820
2-(4-Fluorophenoxy)propanohydrazide
2-(4-Fluorophenoxy)propanohydrazide

Hazard: Irritant

Formula: C₉H₁₁FN₂O₂
MDL Number: MFCD01930249
CAS Number: 590356-83-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR