2-(2-Nitrophenoxy)propanohydrazide

Supplier: Matrix Scientific

029764-500MG
101828-810EA 109.25 USD
101828-810
2-(2-Nitrophenoxy)propanohydrazide
2-(2-Nitrophenoxy)propanohydrazide

Hazard: Irritant

Formula: C₉H₁₁N₃O₄
MDL Number: MFCD02254115
CAS Number: 203741-59-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR