2-(4-Nitrophenoxy)propanohydrazide

Supplier: Matrix Scientific

029763-500MG
101828-808EA 146.15 USD
101828-808
2-(4-Nitrophenoxy)propanohydrazide
2-(4-Nitrophenoxy)propanohydrazide

Hazard: Irritant

Formula: C₉H₁₁N₃O₄
MDL Number: MFCD03423133
CAS Number: 203741-61-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR