2-(4-tert-Butylphenoxy)propanohydrazide

Supplier: Matrix Scientific

029760-500MG
101828-802EA 109.25 USD
101828-802
2-(4-tert-Butylphenoxy)propanohydrazide
2-(4-tert-Butylphenoxy)propanohydrazide

Hazard: Irritant

Formula: C₁₃H₂₀N₂O₂
MDL Number: MFCD02269953
CAS Number: 125096-56-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR