2-(4-Isopropylphenoxy)propanohydrazide

Supplier: Matrix Scientific

029759-500MG
101828-800EA 146.15 USD
101828-800
2-(4-Isopropylphenoxy)propanohydrazide
2-(4-Isopropylphenoxy)propanohydrazide

Hazard: Irritant

Formula: C₁₂H₁₈N₂O₂
MDL Number: MFCD03423132
CAS Number: 667412-84-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR