2-(3-Isobutoxyphenyl)quinoline-4-carbohydrazide

Supplier: Matrix Scientific

029758-500MG
101828-798EA 290.77 USD
101828-798
2-(3-Isobutoxyphenyl)quinoline-4-carbohydrazide
2-(3-Isobutoxyphenyl)quinoline-4-carbohydrazide

Hazard: Irritant

Formula: C₂₀H₂₁N₃O₂
MDL Number: MFCD02375092
CAS Number: 524733-57-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR