2-(4-Ethylphenoxy)propanohydrazide

Supplier: Matrix Scientific

029757-500MG
101828-796EA 146.15 USD
101828-796
2-(4-Ethylphenoxy)propanohydrazide
2-(4-Ethylphenoxy)propanohydrazide

Hazard: Irritant

Formula: C₁₁H₁₆N₂O₂
MDL Number: MFCD03423131
CAS Number: 669705-42-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR