2-(2-Chlorophenoxy)propanohydrazide

Supplier: Matrix Scientific

029756-500MG
101828-794EA 146.15 USD
101828-794
2-(2-Chlorophenoxy)propanohydrazide
2-(2-Chlorophenoxy)propanohydrazide

Hazard: Irritant

Formula: C₉H₁₁ClN₂O₂
MDL Number: MFCD01930247
CAS Number: 52094-94-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR