2-(4-Bromophenoxy)propanohydrazide
Supplier: Matrix Scientific
029754-500MG
101828-790EA
146.15
USD
101828-790
2-(4-Bromophenoxy)propanohydrazide
2-(4-Bromophenoxy)propanohydrazide
Hazard: Irritant
Formula:
C₉H₁₁BrN₂O₂ |
MDL Number:
MFCD00553740 CAS Number: 325778-59-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...