2-(2,3-Dimethylphenoxy)propanohydrazide

Supplier: Matrix Scientific

029753-500MG
101828-788EA 146.15 USD
101828-788
2-(2,3-Dimethylphenoxy)propanohydrazide
2-(2,3-Dimethylphenoxy)propanohydrazide

Hazard: Irritant

Formula: C₁₁H₁₆N₂O₂
MDL Number: MFCD03423129
CAS Number: 142835-36-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR