2-(3,5-Dimethylphenoxy)propanohydrazide

Supplier: Matrix Scientific

029752-500MG
101828-786EA 109.25 USD
101828-786
2-(3,5-Dimethylphenoxy)propanohydrazide
2-(3,5-Dimethylphenoxy)propanohydrazide

Hazard: Irritant

Formula: C₁₁H₁₆N₂O₂
MDL Number: MFCD01567458
CAS Number: 587852-62-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR