2-(4-Methylphenoxy)propanohydrazide

Supplier: Matrix Scientific

029751-500MG
101828-784EA 146.15 USD
101828-784
2-(4-Methylphenoxy)propanohydrazide
2-(4-Methylphenoxy)propanohydrazide

Hazard: Irritant

Formula: C₁₀H₁₄N₂O₂
MDL Number: MFCD02269956
CAS Number: 83798-16-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR