2-(3-Methylphenoxy)propanohydrazide

Supplier: Matrix Scientific

029750-500MG
101828-782EA 146.15 USD
101828-782
2-(3-Methylphenoxy)propanohydrazide
2-(3-Methylphenoxy)propanohydrazide

Hazard: Irritant

Formula: C₁₀H₁₄N₂O₂
MDL Number: MFCD01695446
CAS Number: 90330-07-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR