2-(2-Methylphenoxy)propanohydrazide

Supplier: Matrix Scientific

029749-500MG
101828-780EA 146.15 USD
101828-780
2-(2-Methylphenoxy)propanohydrazide
2-(2-Methylphenoxy)propanohydrazide

Hazard: Irritant

Formula: C₁₀H₁₄N₂O₂
MDL Number: MFCD03423128
CAS Number: 86098-42-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR