2-(3-Isopropoxyphenyl)quinoline-4-carbohydrazide

Supplier: Matrix Scientific

029748-500MG
101828-778EA 290.77 USD
101828-778
2-(3-Isopropoxyphenyl)quinoline-4-carbohydrazide
2-(3-Isopropoxyphenyl)quinoline-4-carbohydrazide

Hazard: Irritant

Formula: C₁₉H₁₉N₃O₂
MDL Number: MFCD02608591
CAS Number: 524733-45-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR