2-(1,3-Benzodioxol-5-yloxy)propanohydrazide

Supplier: Matrix Scientific

029747-500MG
101828-776EA 146.15 USD
101828-776
2-(1,3-Benzodioxol-5-yloxy)propanohydrazide
2-(1,3-Benzodioxol-5-yloxy)propanohydrazide

Hazard: Irritant

Formula: C₁₀H₁₂N₂O₄
MDL Number: MFCD03423127
CAS Number: 588679-99-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR