2-(3-Methyl-4-nitrophenoxy)propanohydrazide

Supplier: Matrix Scientific

029746-500MG
101828-774EA 146.15 USD
101828-774
2-(3-Methyl-4-nitrophenoxy)propanohydrazide
2-(3-Methyl-4-nitrophenoxy)propanohydrazide

Hazard: Irritant

Formula: C₁₀H₁₃N₃O₄
MDL Number: MFCD03423126
CAS Number: 588681-42-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR