2-(2,4-Difluorophenoxy)propanohydrazide

Supplier: Matrix Scientific

029744-500MG
101828-770EA 109.25 USD
101828-770
2-(2,4-Difluorophenoxy)propanohydrazide
2-(2,4-Difluorophenoxy)propanohydrazide

Hazard: Irritant

Formula: C₉H₁₀F₂N₂O₂
MDL Number: MFCD03423123
CAS Number: 588678-32-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR