2-[3-(Trifluoromethyl)phenoxy]propanohydrazide

Supplier: Matrix Scientific

029743-500MG
101828-768EA 146.15 USD
101828-768
2-[3-(Trifluoromethyl)phenoxy]propanohydrazide
2-[3-(Trifluoromethyl)phenoxy]propanohydrazide

Hazard: Irritant

Formula: C₁₀H₁₁F₃N₂O₂
MDL Number: MFCD03423122
CAS Number: 667413-01-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR