2-(4-Bromo-2-chlorophenoxy)propanohydrazide

Supplier: Matrix Scientific

029742-500MG
101828-766EA 146.15 USD
101828-766
2-(4-Bromo-2-chlorophenoxy)propanohydrazide
2-(4-Bromo-2-chlorophenoxy)propanohydrazide

Hazard: Irritant

Formula: C₉H₁₀BrClN₂O₂
MDL Number: MFCD03423121
CAS Number: 588679-51-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR