2-(2-Chloro-5-methylphenoxy)propanohydrazide

Supplier: Matrix Scientific

029741-500MG
101828-764EA 146.15 USD
101828-764
2-(2-Chloro-5-methylphenoxy)propanohydrazide
2-(2-Chloro-5-methylphenoxy)propanohydrazide

Hazard: Irritant

Formula: C₁₀H₁₃ClN₂O₂
MDL Number: MFCD03423120
CAS Number: 588680-01-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR