2-Phenoxypropanohydrazide

Supplier: Matrix Scientific

029740-500MG
101828-762EA 146.15 USD
101828-762
2-Phenoxypropanohydrazide
2-Phenoxypropanohydrazide

Hazard: Irritant

Formula: C₉H₁₂N₂O₂
MDL Number: MFCD01463172
CAS Number: 52094-92-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR